Anti-Krt18 Rabbit Recombinant Polyclonal Antibody

Supplier: AFG Bioscience
A27176-100UG A27176-50UG
77741-867EA 330 USD
77741-867 77858-735
Anti-Krt18 Rabbit Recombinant Polyclonal Antibody
Antibodies
Recommended dilution: WB: 1:500 to 1:5000, IHC: 1:20 to 1:200

Type:
Antigen: Krt18
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse;Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR