Anti-IRF7 Rabbit Recombinant Polyclonal Antibody

Supplier: AFG Bioscience
A26449-100UG A26449-50UG
77741-196EA 330 USD
77741-196 77858-202
Anti-IRF7 Rabbit Recombinant Polyclonal Antibody
Antibodies
Recommended dilution: IHC:1:20 to 1:200, IF:1:50 to 1:200

Type:
Antigen: IRF7
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR