Anti-IRAK3 Rabbit Recombinant Polyclonal Antibody

Supplier: AFG Bioscience
A26414-100UL A26414-50UL
77741-166EA 330 USD
77741-166 77858-173
Anti-IRAK3 Rabbit Recombinant Polyclonal Antibody
Antibodies
Type:
Antigen: IER2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR