Anti-HIST1H3A (di-methyl-K4) Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A25075-100UL A25075-50UL
77739-976EA 350 USD
77739-976 77857-290
Anti-HIST1H3A (di-methyl-K4) Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: ICC:1:20 to 1:200, IF:1:50 to 1:200

Type:
Antigen: HIST1H3A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR