Anti-HAP1 Rabbit Recombinant Polyclonal Antibody (Biotin)

Supplier: AFG Bioscience
A24340-100UL A24340-50UL
77739-413EA 330 USD
77739-413 77856-716
Anti-HAP1 Rabbit Recombinant Polyclonal Antibody (Biotin)
Antibodies
Type:
Antigen: HAP1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR