Anti-GPR1 Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))

Supplier: AFG Bioscience
A23666-100UG A23666-50UG
77738-772EA 330 USD
77738-772 77856-329
Anti-GPR1 Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antibodies
Type:
Antigen: GPR1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR