Anti-GPM6A Rabbit Recombinant Polyclonal Antibody

Supplier: AFG Bioscience
A23654-100UG A23654-50UG
77738-760EA 330 USD
77738-760 77856-320
Anti-GPM6A Rabbit Recombinant Polyclonal Antibody
Antibodies
Recommended dilution: WB: 1:500 to 1:5000, IF: 1:50 to 1:200

Type:
Antigen: GPM6A
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR