Anti-GLP2R Rabbit Recombinant Polyclonal Antibody

Supplier: AFG Bioscience
A23288-100UL A23288-50UL
77738-417EA 330 USD
77738-417 77856-101
Anti-GLP2R Rabbit Recombinant Polyclonal Antibody
Antibodies
Recommended dilution: IHC: 1:20 to 1:200

Type:
Antigen: GLP2R
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR