Anti-GBA Rabbit Recombinant Polyclonal Antibody (Biotin)

Supplier: AFG Bioscience
A22864-100UG A22864-50UG
77738-023EA 330 USD
77738-023 77855-832
Anti-GBA Rabbit Recombinant Polyclonal Antibody (Biotin)
Antibodies
Type:
Antigen: GBA
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR