Anti-EMD Rabbit Recombinant Polyclonal Antibody

Supplier: AFG Bioscience
A20535-50UG
77854-693EA 225 USD
77854-693
Anti-EMD Rabbit Recombinant Polyclonal Antibody
Antibodies
Recommended dilution: WB:1:500 to 1:5000, IHC:1:20 to 1:200, IF:1:50 to 1:200, IP:1:200 to 1:2000

Type:
Antigen: EMD
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR