Anti-RPS25 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Supplier: AFG Bioscience
A18895-50UG
77853-948EA 225 USD
77853-948
Anti-RPS25 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antibodies
Type:
Antigen: RPS25
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR