Anti-CSF1R Rabbit Recombinant Polyclonal Antibody

Supplier: AFG Bioscience
A18268-50UG
77853-642EA 225 USD
77853-642
Anti-CSF1R Rabbit Recombinant Polyclonal Antibody
Antibodies
Type:
Antigen: CSF1R
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Felis silvestris catus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR