Anti-COL9A1 Rabbit Recombinant Polyclonal Antibody

Supplier: AFG Bioscience
A17828-50UL
77853-405EA 225 USD
77853-405
Anti-COL9A1 Rabbit Recombinant Polyclonal Antibody
Antibodies

Recommended dilution: WB:1:1000 to 1:5000



Type:
Antigen: COL9A1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR