Anti-CD5 Rabbit Recombinant Polyclonal Antibody

Supplier: AFG Bioscience
A16582-50UG
77852-691EA 225 USD
77852-691
Anti-CD5 Rabbit Recombinant Polyclonal Antibody
Antibodies
Recommended dilution: IF: 1:50 to 1:200

Type:
Antigen: CD5
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR