Anti-AMACR Rabbit Recombinant Polyclonal Antibody (Biotinylated)

Supplier: AFG Bioscience
A12513-50UG
77851-098EA 225 USD
77851-098
Anti-AMACR Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Antibodies
Type:
Antigen: AMACR
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR