Anti-ALOX12 Rabbit Recombinant Polyclonal Antibody

Supplier: AFG Bioscience
A12457-50UG
77851-066EA 225 USD
77851-066
Anti-ALOX12 Rabbit Recombinant Polyclonal Antibody
Antibodies
Recommended dilution: WB: 1:500 to 1:5000, IHC: 1:500 to 1:1000, IF: 1:50 to 1:200

Type:
Antigen: IER2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotinylated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR