Anti-Aif1 Rabbit Recombinant polyclonal Antibody (Biotinylated)

Supplier: AFG Bioscience
A12223-50UG
77850-932EA 225 USD
77850-932
Anti-Aif1 Rabbit Recombinant polyclonal Antibody (Biotinylated)
Antibodies
0

Type:
Antigen: Aif1
Clonality: Recombinant polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR