Anti-AGGF1 Rabbit Recombinant Polyclonal Antibody

Supplier: AFG Bioscience
A12126-50UL
77850-868EA 225 USD
77850-868
Anti-AGGF1 Rabbit Recombinant Polyclonal Antibody
Antibodies
Recommended dilution: IHC:1:20 to 1:200

Type:
Antigen: IER2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR