Anti-IRF3 (phospho-Ser386) HEK293F cells Recombinant Monoclonal Antibody [clone: 4H10]

Supplier: AFG Bioscience
A74865-50UL
77850-287EA 235 USD
77850-287
Anti-IRF3 (phospho-Ser386) HEK293F cells Recombinant Monoclonal Antibody [clone: 4H10]
Antibodies
Recommended dilution: WB: 1:500 to 1:5000

Type:
Antigen: IRF3
Clonality: Recombinant Monoclonal
Clone: 4H10
Conjugation: Unconjugated
Epitope:
Host: HEK293F cells
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR