Anti-GPX1 HEK293F cells Recombinant Monoclonal Antibody [clone: 3G1]

Supplier: AFG Bioscience
A74812-100UL A74812-50UL
77850-223EA 375 USD
77850-223 77850-224
Anti-GPX1 HEK293F cells Recombinant Monoclonal Antibody [clone: 3G1]
Antibodies
Type:
Antigen: GPX1
Clonality: Recombinant Monoclonal
Clone: 3G1
Conjugation: Unconjugated
Epitope:
Host: HEK293F cells
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR