Anti-CDC25B HEK293F cells Recombinant Monoclonal Antibody [clone: 3D6]

Supplier: AFG Bioscience
A74779-100UL A74779-50UL
77850-183EA 375 USD
77850-183 77850-184
Anti-CDC25B HEK293F cells Recombinant Monoclonal Antibody [clone: 3D6]
Antibodies
Type:
Antigen: CDC25B
Clonality: Recombinant Monoclonal
Clone: 3D6
Conjugation: Unconjugated
Epitope:
Host: HEK293F cells
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR