Anti-CD97 HEK293F cells Recombinant Monoclonal Antibody [clone: 3A7]

Supplier: AFG Bioscience
A74777-50UL
77850-181EA 235 USD
77850-181
Anti-CD97 HEK293F cells Recombinant Monoclonal Antibody [clone: 3A7]
Antibodies
Recommended dilution: IHC:1:50 to 1:500, IF:1:30 to 1:200

Type:
Antigen: IER2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR