Anti-CD300LF Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A74525-100UL
77849-765EA 330 USD
77849-765
Anti-CD300LF Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: WB: 1:500 to 1:3000

Type:
Antigen: IER2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotinylated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR