Anti-CD177 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A74410-100UL A74410-50UL
77849-577EA 330 USD
77849-577 77849-578
Anti-CD177 Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: ELISA: 1:1000 to 1:2000, IHC: 1:25 to 1:100

Type:
Antigen: CD177
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR