Anti-SYNCRIP Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A74320-100UL A74320-50UL
77849-427EA 330 USD
77849-427 77849-428
Anti-SYNCRIP Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: ELISA: 1:2000 to 1:5000, IHC: 1:25 to 1:100

Type:
Antigen: SYNCRIP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR