Anti-TRAF3IP1 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A74144-100UL A74144-50UL
77849-136EA 330 USD
77849-136 77849-137
Anti-TRAF3IP1 Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: ELISA: 1:2000 to 1:10000, WB: 1:200 to 1:1000, IHC: 1:50 to 1:200

Type:
Antigen: TRAF3IP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR