Anti-SH2D3A Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A74135-100UL A74135-50UL
77849-120EA 330 USD
77849-120 77849-121
Anti-SH2D3A Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: ELISA: 1:1000 to 1:2000, IHC: 1:25 to 1:100

Type:
Antigen: SH2D3A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR