Anti-SMURF2 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A74094-100UL A74094-50UL
77849-046EA 330 USD
77849-046 77849-047
Anti-SMURF2 Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: ELISA: 1:1000 to 1:2000, IHC: 1:25 to 1:100

Type:
Antigen: SMURF2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR