Anti-SPATA5L1 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A73982-100UL A73982-50UL
77848-857EA 330 USD
77848-857 77848-858
Anti-SPATA5L1 Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: ELISA: 1:1000 to 1:2000, WB: 1:200 to 1:1000, IHC: 1:25 to 1:100

Type:
Antigen: SPATA5L1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR