Anti-SIRT6 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A73946-100UL A73946-50UL
77848-797EA 330 USD
77848-797 77848-798
Anti-SIRT6 Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: ELISA: 1:2000 to 1:5000, WB: 1:500 to 1:2000

Type:
Antigen: SIRT6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR