Anti-KCNQ1 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A73627-100UL A73627-50UL
77848-280EA 330 USD
77848-280 77848-281
Anti-KCNQ1 Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: ELISA: 1:1000 to 1:2000, WB: 1:200 to 1:1000, IHC: 1:50 to 1:200

Type:
Antigen: KCNQ1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR