Anti-CBL Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A73401-100UL
77847-898EA 330 USD
77847-898
Anti-CBL Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: WB: 1:500 to 1:1000, IHC: 1:50 to 1:200

Type:
Antigen: CBL
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR