Anti-IL3RA Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A73254-50UL A73254-100UL
77847-654EA 225 USD
77847-654 77847-653
Anti-IL3RA Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: ELISA: 1:2000 to 1:5000, WB: 1:500 to 1:2000, IHC: 1:25 to 1:100

Type:
Antigen: IL3RA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR