Anti-SDC3 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A73247-100UL A73247-50UL
77847-642EA 330 USD
77847-642 77847-643
Anti-SDC3 Rabbit Polyclonal Antibody
Antibodies

Recommended dilution: WB:1:1000 to 1:5000



Type:
Antigen: SDC3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR