Anti-SNCB Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A73210-100UL
77847-581EA 330 USD
77847-581
Anti-SNCB Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: WB: 1:500 to 1:3000, IHC: 1:50 to 1:100, IF: 1:100 to 1:500

Type:
Antigen: SNCB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR