Anti-FZD4 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A73019-100UL A73019-50UL
77847-264EA 330 USD
77847-264 77847-265
Anti-FZD4 Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: ELISA:1:1000 to 1:2000, WB:1:200 to 1:1000

Type:
Antigen: FZD4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR