Anti-RALY Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))

Supplier: AFG Bioscience
A71714-100UG A71714-50UG
77844-897EA 330 USD
77844-897 77844-898
Anti-RALY Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antibodies
Type:
Antigen: RALY
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR