Anti-USP18 Rabbit Recombinant polyclonal Antibody (HRP (Horseradish Peroxidase))

Supplier: AFG Bioscience
A71404-100UG A71404-50UG
77844-315EA 330 USD
77844-315 77844-316
Anti-USP18 Rabbit Recombinant polyclonal Antibody (HRP (Horseradish Peroxidase))
Antibodies
Type:
Antigen: USP18
Clonality: Recombinant polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR