Anti-MACF1 Rabbit Recombinant polyclonal Antibody (Biotinylated)

Supplier: AFG Bioscience
A71314-100UL A71314-50UL
77844-146EA 330 USD
77844-146 77844-147
Anti-MACF1 Rabbit Recombinant polyclonal Antibody (Biotinylated)
Antibodies
Type:
Antigen: MACF1
Clonality: Recombinant polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR