Anti-EGFL7 Rabbit Recombinant Polyclonal Antibody

Supplier: AFG Bioscience
A71157-100UG A71157-50UG
77843-851EA 330 USD
77843-851 77843-852
Anti-EGFL7 Rabbit Recombinant Polyclonal Antibody
Antibodies
Recommended dilution: IHC: 1:200 to 1:500

Type:
Antigen: EGFL7
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR