Anti-FAIM Rabbit Recombinant Polyclonal Antibody

Supplier: AFG Bioscience
A70990-100UG A70990-50UG
77843-542EA 330 USD
77843-542 77843-543
Anti-FAIM Rabbit Recombinant Polyclonal Antibody
Antibodies
Recommended dilution: WB: 1:500 to 1:5000, IHC: 1:20 to 1:200, IF: 1:50 to 1:200

Type:
Antigen: FAIM
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR