Anti-BRD9 Rabbit Recombinant polyclonal Antibody

Supplier: AFG Bioscience
A70819-100UL A70819-50UL
77843-218EA 330 USD
77843-218 77843-219
Anti-BRD9 Rabbit Recombinant polyclonal Antibody
Antibodies
Recommended dilution: IHC:1:20-1:200, IF:1:50-1:200

Type:
Antigen: BRD9
Clonality: Recombinant polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR