Anti-MAP1LC3B Rabbit Recombinant Polyclonal Antibody

Supplier: AFG Bioscience
A70723-100UG A70723-50UG
77843-040EA 330 USD
77843-040 77843-041
Anti-MAP1LC3B Rabbit Recombinant Polyclonal Antibody
Antibodies
Recommended dilution: IHC:1:20 to 1:200

Type:
Antigen: MAP1LC3B
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR