Anti-SMC4 Rabbit Recombinant Polyclonal Antibody (HRP)

Supplier: AFG Bioscience
A70384-100UG A70384-50UG
77842-396EA 330 USD
77842-396 77842-397
Anti-SMC4 Rabbit Recombinant Polyclonal Antibody (HRP)
Antibodies
Type:
Antigen: SMC4
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR