Anti-TP53AIP1 Rabbit Recombinant Polyclonal Antibody

Supplier: AFG Bioscience
A70289-100UL A70289-50UL
77842-216EA 330 USD
77842-216 77842-217
Anti-TP53AIP1 Rabbit Recombinant Polyclonal Antibody
Antibodies
Recommended dilution: IHC: 1:20 to 1:200

Type:
Antigen: TP53AIP1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR