Anti-RIBC2 Rabbit Recombinant Polyclonal Antibody

Supplier: AFG Bioscience
A70190-50UG
77842-034EA 225 USD
77842-034
Anti-RIBC2 Rabbit Recombinant Polyclonal Antibody
Antibodies
Recommended dilution: WB: 1:1000 to 1:5000, IHC: 1:20 to 1:200

Type:
Antigen: RIBC2
Clonality: Recombinant Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR