Anti-FUZ Rabbit Recombinant Polyclonal Antibody (FITC)

Supplier: AFG Bioscience
A70002-100UG A70002-50UG
77841-672EA 330 USD
77841-672 77841-673
Anti-FUZ Rabbit Recombinant Polyclonal Antibody (FITC)
Antibodies
Type:
Antigen: FUZ
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR