Anti-NRIP2 Rabbit Recombinant Polyclonal Antibody (FITC)

Supplier: AFG Bioscience
A69236-100UG A69236-50UG
77840-220EA 330 USD
77840-220 77840-221
Anti-NRIP2 Rabbit Recombinant Polyclonal Antibody (FITC)
Antibodies
Type:
Antigen: NRIP2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR