Anti-PDP1 Rabbit Recombinant Polyclonal Antibody (HRP)

Supplier: AFG Bioscience
A69155-50UG A69155-100UG
77840-068EA 225 USD
77840-068 77840-067
Anti-PDP1 Rabbit Recombinant Polyclonal Antibody (HRP)
Antibodies
Type:
Antigen: PDP1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR