Anti-PRDM10 Rabbit Recombinant Polyclonal Antibody (HRP)

Supplier: AFG Bioscience
A68998-100UG A68998-50UG
77839-766EA 330 USD
77839-766 77839-767
Anti-PRDM10 Rabbit Recombinant Polyclonal Antibody (HRP)
Antibodies
Type:
Antigen: PRDM10
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR